missing translation for 'onlineSavingsMsg'
Learn More

SGE™ GC Capillary Columns: Cyanopropyl Polysilphenylene Siloxane

Range of columns that feature high levels of robustness, reproducibility and performance

Brand:  SGE™ 054620

 View more versions of this product

Product Code. 10451041

  • £687.00 / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Range of columns that feature high levels of robustness, reproducibility and performance

  • Low bleed; high inertness for most demanding analyses
  • High tolerance level to adverse conditions and samples
  • Manufactured to exact specifications, ensuring column-to-column reproducibilty

  • forte BPX-70 (70% Cyanopropyl Polysilphenylene-siloxane)—MS-premium, low bleed column with a polarity that has been designed for Fatty Acid Methyl ester (FAME) analysis
Specifications

Specifications

260°C
70% Cyanopropyl Polysilphenylene Siloxane
0.5 μm
0.5 μm
BPX70
0.53 mm
GC/MS, cis/trans Isomer Analysis
50°C to 250°C
30 m
Polar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.