missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ GC Deactivated Inlet Liner Split Quartz Wool For Agilent 7890
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09200101
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent instruments
Specifications
4 mm | |
78.5 mm | |
Inlet Liner with Quartz Wool |