Learn More
Trajan™ Sealing Ring for GC Ferrule
Graphite sealing ring for use with GC Ferrules
Brand: Trajan™ 0726520
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- For use with GC Ferrules
- 100% Graphite Sealing ring for 1/4 in. fittings
- Supplied in packs of 10
Specifications
1/16 in. | |
1.7 mm | |
450°C | |
3.2 mm |
1.59 mm | |
100% Graphite | |
Sealing Ring |