missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ GC inlet liner packed column for Agilent packed columns 1.8mm I.D., 3mm O.D., 92mm length SGE
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092235
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent instruments
Specifications
1.8 mm | |
92 mm | |
1.8 mm I.D. Packed Column Liner | |
3 mm |