Learn More
Thermo Scientific™ GC Installation Kit
Add a GC system to your lab bench with the Thermo Scientific™ GC Installation Kit. The required tubing and fittings are conveniently supplied in this kit.
Brand:  Thermo Scientific™ 60180-888
583.04 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10589165
  • £587.93 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Add a GC system to your lab bench with the Thermo Scientific™ GC Installation Kit. The required tubing and fittings are conveniently supplied in this kit.
Includes:
Tubing cutter; 1/8 x 1/4in. reamer; 7/16in. wrench; 1/2in. wrench; 1/8in. brass tees, 4; 1/8in. brass nuts, 10; Brass front and back ferrules, 10; 15.2m instrument-grade, cleaned 1/8in. copper tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GC Installation Kit
GC
Gas Chromatography Systems
GC Installation Kit
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GC Installation Kit > GC Installation Kit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.