Learn More
Thermo Scientific™ GC Tool Kit for Agilent Instruments
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
Brand: Thermo Scientific™ 60180-786
663.39 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
- Replacements included perform like original instrument parts
- Parts list included for easy reordering
Includes:
Capillary installation gauge, injector wrench, septum nut removal tool, open-end wrenches in 7/16 x 1/2in. and 1/2 x 9/16in. sizes.
Specifications
Capillary Tool Kit | |
GC Tool Kit for Agilent Instruments | |
GC | |
Agilent GC-MS Instruments |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.