missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments.
Brand: Epredia™ MS-1170-RQ
This item is not returnable.
View return policy
Description
Gross cystic disease of the breast is benign premenopausal disorder in which cysts are a predominant pathological lesion. These cysts appear to be formed from excessive apocrine cystic secretions. This fluid is composed of several glycoproteins including a unique 15 kDa monomer protein, Gross Cystic Disease Fluid Protein-15 (GCDFP15). Cytosolic analysis of normal tissue specimens from all major organs has demonstrated GCDFP15 in apocrine epithelia, lacrimal, ceruminous and Moll's glands and in numerous serous cells of the submandibular, tracheal, bronchial, sublingual and minor salivary glands. GCDFP15 and prostate specific antigen are co-expressed in androgen receptor-positive breast tumours.Host Species: Mouse
Clone: 23A3
Isotype: IgG2a/κ
Species Reactivity: Human and Rat. Others not known.
Immunogen: Recombinant protein encoding the excreted domain of human GCDFP-15
Molecular Weight: 15 kDa
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding the excreted domain of human GCDFP15 | |
IVD | |
Primary |
Immunohistochemistry (Paraffin) | |
23A3 | |
Mouse | |
12 mL | |
Cancer and Tumor Biology | |
Ready-to-Use |