missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments.

Brand:  Epredia™ MS-1170-S1

 View more versions of this product

Product Code. 12699196

  • £340.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Gross cystic disease of the breast is benign premenopausal disorder in which cysts are a predominant pathological lesion. These cysts appear to be formed from excessive apocrine cystic secretions. This fluid is composed of several glycoproteins including a unique 15 kDa monomer protein, Gross Cystic Disease Fluid Protein-15 (GCDFP15). Cytosolic analysis of normal tissue specimens from all major organs has demonstrated GCDFP15 in apocrine epithelia, lacrimal, ceruminous and Moll's glands and in numerous serous cells of the submandibular, tracheal, bronchial, sublingual and minor salivary glands. GCDFP15 and prostate specific antigen are co-expressed in androgen receptor-positive breast tumours.

Host Species: Mouse

Clone: 23A3

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not known.

Immunogen: Recombinant protein encoding the excreted domain of human GCDFP-15

Molecular Weight: 15 kDa

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1
Monoclonal
Unconjugated
Recombinant protein encoding the excreted domain of human GCDFP15
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
23A3
Mouse
500 μL
Cancer and Tumor Biology
Human, Rat
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers