missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCDFP-15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121963
This item is not returnable.
View return policy
Description
GCDFP-15 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Gross cystic disease of the breast is benign premenopausal disorder in which cysts are a predominant pathological lesion. These cysts appear to be formed from excessive apocrine cystic secretions. This fluid is composed of several glycoproteins including a unique 15kDa monomer protein, Gross Cystic Disease Fluid Protein-15 (GCDFP15). Cytosolic analysis of normal tissue specimens from all major organs has demonstrated GCDFP15 in apocrine epithelia, lacrimal, ceruminous and Moll's glands and in numerous serous cells of the submandibular, tracheal, bronchial, sublingual and minor salivary glands. GCDFP15 and prostate specific antigen are co-expressed in androgen receptor-positive breast tumours.
Specifications
| GCDFP-15 | |
| Polyclonal | |
| Unconjugated | |
| PIP | |
| 14 kDa submandibular gland protein; gcdfp 15; GCDFP15; GCDFP-15; gp17; GPIP4; Gross cystic disease fluid protein 15; HGNC:8993; mSMGP; PIP; prolactin induced protein; prolactin-induced protein; Prolactin-inducible protein; prolactin-inducible protein homolog; SABP; secretory actin-binding protein; SMGP | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 64673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O70417 | |
| PIP | |
| Recombinant protein Prolactin Induced Protein. The antigen corresponds to amino acid range 41-139 of the target protein. | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction