missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDNF Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHGDNFX5

Additional Details : Weight : 0.33820kg

 View more versions of this product

Product Code. 17195449

  • £2320.00 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket

Description

Description

The Human Glial-Derived Neurotrophic Factor (Hu GDNF) ELISA quantitates Hu GDNF in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu GDNF. Principle of the method The Human GDNF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.
Specifications

Specifications

P39905
4 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
2668
<12%
Wash Buffer, SAV-HRP, Adhesive Plate Covers, Assay Diluent Concentrate, Standard, Biotinylated Detection Antibody, Stop Solution, Pre-coated 96 well plates, Chromogen
5 x 96 Tests
Neurobiology
2°C to 8°C
Human
4 hr. 45 min.
2.74 pg/mL to 2000 pg/mL
4 pg/mL to 2000 pg/mL
Biotin
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
ATF,ATF1,ATF2,HFB1-GDNF,HSCR3
<10%
HRP
RUO
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L
ATF1, ATF2, HFB1-GDNF, HSCR3
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers