missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Geared travel trolley, beam width 50/220mm, 500kg capacity
Brand: Manutan A338488
This item is not returnable.
View return policy
Description
Position large loads with either manual or powered hoisting equipment. Excellent low rolling resistance due to machined steel wheels, mounted on pre-lubricated, encapsulated ball bearings. Adjustable to fit a wide range of beam widths and profiles. Clevis bar ensures centred positioning of the hoist in the clevis, eliminating left or right creeping.- Load capacity: 500kg
- Track width: 50mm-220mm
- Dimensions (H x W x D): 300mm x 250mm x 330mm