Learn More
Cytiva Gel Casters for MiniVE and SE250, 260 Vertical Units
For MiniVE, SE250 and SE260 vertical electrophoresis units
Brand: Cytiva SE215
This item is not returnable.
View return policy
Description
Cast up to 10 minigels at once.
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System | |
Gel Caster | |
10 Notched Alumina plates, 20 rectangular Glass plates, 2 red clamps, 100 sheets Wax Paper, space-saver plate, 5 filler sheets, set of filler plug and spacer-mate assembly template | |
4 to 10 |