Learn More
Cytiva GelBond PAG Gel Film
Cytiva™ GelBond PAG Gel Film is a high density homogenous medium.
Brand:  Cytiva 80112936
 View more versions of this product
Product Code. 10086234
  • £536.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Cytiva™ GelBond Film is a transparent, flexible polyester film designed to support gels. Gels cast on GelBond Film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures.
  • GelBond Film is made from thin polyester (0.2mm). Pack of 50 films, 124 x 258mm.
  • High-density homogenous medium
Each film has one hydrophilic side and one hydrophobic side. The gel must be cast on the hydrophilic side, which is the side where a water droplet will spread out.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Colorless
Polyacrylamide gels
10.1 in.
5.0 in.
Gel Film
50 Pack
258 mm
124 mm