missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GelBond PAG Gel Film

Cytiva™ GelBond PAG Gel Film is a high density homogenous medium.

Brand:  Cytiva 80112933

245.79 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10575785

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cytiva™ GelBond Film is a transparent, flexible polyester film designed to support agarose gels. Gels cast on GelBond Film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures. 
  • GelBond Film is made from thin polyester (0.2mm). Pack of 50 films, 124 x 258mm.
  • High-density homogenous medium
Each film has one hydrophilic side and one hydrophobic side. The gel must be cast on the hydrophilic side, which is the side where a water droplet will spread out.  
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
Solid
50 Pack
94 mm
84 mm
Gel Film
Agarose gels
3.7 in.
3.3 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.