missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GelCode™ Blue Stain Reagent
Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
3.5 L
Unit Size:
500mL
Each
Description
GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection. This unique reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process.
GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.
Features include:
• No methanol-acetic acid destaining required
• Fast, one-step, one-hour staining, ready-to-use stain reagent; band development visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
• Water wash increases staining sensitivity; optional step provides crystal-clear gel background and makes weakly stained bands more visible
GelCode Blue Stain Reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process. Because the destaining step does not fix the protein, GelCode Blue Stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.
Features include:
• No methanol-acetic acid destaining required
• Fast, one-step, one-hour staining, ready-to-use stain reagent; band development visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
• Water wash increases staining sensitivity; optional step provides crystal-clear gel background and makes weakly stained bands more visible
GelCode Blue Stain Reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process. Because the destaining step does not fix the protein, GelCode Blue Stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Detection Location | In-Blot Detection, In-Gel Detection |
| Color | Blue |
| Detection Method | Colorimetric |
| Label or Dye | Coomassie |
| Product Type | Protein Gel Stain Reagent |
| Product Line | GelCode |
| Quantity | 3.5 L |
| Target Molecule | Protein |
Safety and Handling
Product Identifier
- Gelcode Blue Stain Reagent, 3.5 L
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 2
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H371-May cause damage to organs.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Phosphoric acid, Methyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction