missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GelCode™ Blue Stain Reagent

Product Code. p-4530041 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
3.5 L
Unit Size:
500mL
Each
This item is not returnable. View return policy

Product Code. 10608494

Brand: Thermo Scientific™ 24590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection. This unique reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process.

GelCode Blue Stain Reagent is a ready-to-use protein stain based on colloidal coomassie dye G-250 that provides nanogram-level detection in polyacrylamide gels and for in-blot detection.

Features include:
• No methanol-acetic acid destaining required
• Fast, one-step, one-hour staining, ready-to-use stain reagent; band development visible directly in the staining tray; no background from overnight staining; flexible fixing and washing protocol; no gel shrinkage; stained gel can be dried
• Water wash increases staining sensitivity; optional step provides crystal-clear gel background and makes weakly stained bands more visible

GelCode Blue Stain Reagent stains only protein and allows bands to be viewed directly in the gel during the one-hour gel-staining process. Because the destaining step does not fix the protein, GelCode Blue Stain is compatible with mass spectrometry analysis and N-terminal sequence analysis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Detection Location In-Blot Detection, In-Gel Detection
Color Blue
Detection Method Colorimetric
Label or Dye Coomassie
Product Type Protein Gel Stain Reagent
Product Line GelCode
Quantity 500 mL
Target Molecule Protein
compliance-icons compliance-icons
Product Identifier
  • GelCode Blue Stain Reagent
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methyl alcohol, Phosphoric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.