missing translation for 'onlineSavingsMsg'
Learn More

Gelsolin Monoclonal Antibody (GS-2C4), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA124939

Product Code. 11534832

  • £437.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recommended positive controls: human plasma. The predicted molecular weight is 86 kDa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene binds to the "e;plus"e; ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gelsolin
Monoclonal
3.4 mg/mL
Ascites with 15mM sodium azide
P06396, P20305, Q3SX14
GSN
human plasma gelsolin
RUO
100337841, 106509249, 2934, 396874, 535077
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
GS-2C4
Unconjugated
GSN
Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1
Mouse
100 μL
Primary
Bovine, Human, Porcine, Rabbit
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers