missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
Description
- Routinely used for attachment and maintenance of human embryonic stem cells (hESC)
- Human ESCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support
- Geltrex™ hESC-qualified reduced growth factor basement membrane matrix has been tested for support of hESC growth and pluripotency, therefore minimizing the need to test multiple lots
- Consistent manufacturing processes help ensure a protein concentration averaging 15 mg/mL with very little variation from lot-to-lot
Specifications
Specifications
| Cell Type | Human induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells |
| Classification | Reduced Growth Factor |
| Concentration | 12 to 18 mg/mL |
| Form | Frozen |
| Product Type | Basement Membrane Matrix |
| Serum Level | Serum-free |
| Sterility | Sterile |
| Without Additives | No Phenol Red |
| Product Line | Gibco™ |
| Quantity | 5 mL |
| Show More |
Safety and Handling
Product Identifier
- (2S)-Bornane-10,2-sultam
- MIXTURE LIST-Contains : (+)-10,2-Camphorsultam
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified >
Spot an opportunity for improvement?Share a Content Correction