missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix
A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.
£52.50 - £245.00
Specifications
Cell Type | Human induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells |
---|---|
Classification | Reduced Growth Factor |
Concentration | 12 to 18 mg/mL |
Culture Type | Adherent Cell Culture |
Form | Frozen |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11612149
|
Gibco™
A1413301 |
1 mL |
£52.50
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12063569
|
Gibco™
A1413302 |
5 mL |
£245.00
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Routinely used for attachment and maintenance of human embryonic stem cells (hESC)
- Human ESCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support
- Geltrex™ hESC-qualified reduced growth factor basement membrane matrix has been tested for support of hESC growth and pluripotency, therefore minimizing the need to test multiple lots
- Consistent manufacturing processes help ensure a protein concentration averaging 15 mg/mL with very little variation from lot-to-lot
Specifications
Human induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells | |
12 to 18 mg/mL | |
Frozen | |
Serum-free | |
No Phenol Red | |
Tube(s) | |
Cell Culture Grade | |
Mouse |
Reduced Growth Factor | |
Adherent Cell Culture | |
Basement Membrane Matrix | |
Sterile | |
Low | |
Gibco™ | |
Dry Ice |
Safety and Handling
missing translation for 'shelfLife' : 36 months
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title