missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Gemini X2 BiPolar Electroporation System

All of the benefits of the Gemini X2 Dual Waveform Electroporator, with the addition of bipolar polarity switching which enhances transfection efficiency by preventing transfectant accumulation at a single electrode and enhancing delivery to the target cells or tissue.

Brand:  BTX™ 452062

Product Code. 16614862

  • £15400.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features and Benefits:

  • Square wave and exponential decay waves power in a single unit
  • Multiple pulsing capabilities in both square and exponential decay waveforms (X2 systems)
  • Large 7-inch, easy-to-use touch screen interface
  • Flexible, universal electroporation – transfects cells in 96-well format, vitro, in vivo, in ovo and adherent forms, dependent on accessory
  • Preset protocols – includes the most common eukaryotic and prokaryotic cell types
  • User-defined protocols – unlimited ability to add and modify protocols
  • Safety – displays resistance measurements for each pulse with three layers of arc protection
  • Data management – stores logs of every pulse delivered for QC and troubleshooting
  • Polarity switching enhances transfection efficiency by preventing transfectant accumulation at a single electrode and enhancing delivery to the target cells or tissue

Applications:

  • Bipolar Electroporation
  • CRISPR Transfections
  • Suspension and Adherent Cells
  • In Vivo, In Ovo and In Utero Gene and Drug Delivery
  • Tissue Explants
  • Transformation of Eukaryotic and Prokaryotic Cells
  • High Throughput Electroporation

Supplied with:

  • Gemini X2 Generator
  • Pulse Switcher
  • Power Supply and USB Connector Cable
  • Safety Dome
  • Cuvettes 1 mm, 2 mm, 4 mm (pkg. of 30, 10 each)
  • Cuvette Rack
Specifications

Specifications

Electroporation, CRISPR Transfections, Cell Suspension, Gene and Drug Delivery, Tissue Explants, Cell Transformation
100/240 V
Non-sterile
505 to 3000 V (in 5 V Steps)
Output Cable
Exponential Decay Wave, Square Wave
Electroporation System
Color Touchscreen
All Cell Types
5 to 500 V (in 1 V Steps)
Touchscreen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.