Learn More
Agilent BioTek Gen5 Data Collection and Analysis Software
Gen5 Data Collection and Analysis Software has a contemporary interface which provides easy access to all major functions of the software including reading parameters, plate layout, data reduction, online help, wizards, tutorials and sample protocols.
£291.00 - £16560.00
Specifications
For Use With (Equipment) | Imaging Multi-Mode Readers |
---|---|
Type | Data Collection and Analysis Software |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10186572
|
Agilent BioTek
GEN5 |
Imaging Multi-Mode Readers |
£3630.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15351470
|
Agilent BioTek
GEN5IPLUS |
Imaging Multi-Mode Readers |
£9430.00
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10329742
|
Agilent BioTek
GEN5SECURE |
Imaging Multi-Mode Readers |
£4970.00
Each |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15649408
|
Agilent BioTek
GEN5AL2 |
Imaging Multi-Mode Readers |
£291.00
Each |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16592342
|
Agilent BioTek
GEN5SECUREIPRIME |
Imaging Multi-Mode Readers |
£16560.00
Each |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16552342
|
Agilent BioTek
GEN5AL3 |
Imaging Multi-Mode Readers |
£585.00
Each |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16562342
|
Agilent BioTek
GEN5AL4 |
Imaging Multi-Mode Readers |
£870.00
Each |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16582342
|
Agilent BioTek
GEN5SECUREIPLUS |
Imaging Multi-Mode Readers |
£10770.00
Each |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16572342
|
Agilent BioTek
GEN5IVDIPLUS |
Imaging Multi-Mode Readers |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15386196
|
Agilent BioTek
GEN5IVD |
Imaging Multi-Mode Readers |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Quick launch buttons on the welcome screen provide links to recently used files, pre-programmed protocols and other key functions such as Quick Read and System Test
- StepWise tools allow limitless combinations of procedural steps and data reduction criteria for a wide variety of applications in a logical and clearly defined sequence that is easily viewed on the screen
- Analyze data on a single plate or across multiple plates in an experiment
- Report and PowerExport builders are fast and flexible output tools to produce publication-ready reports using data matrices, statistics tables, protocols and plate information
- For compliance to 21 CFR Part 11, Gen5 Secure provides system, data and protocol protection
- Multiple user definitions and permission levels make it easy to comply with laboratory policies
- Windows XP or Windows 2000 (Professional Editions)
- PentiumIII-Class PC (or compatible) processor (500MHz or higher)
- 512 MB RAM or higher
- 2 GB hard drive space or higher
- XGA resolution at 1024 x 768 or higher
- CD-ROM readable drive
- Keyboard
- Mouse
- Microsoft Internet Explorer v 5.0 or higher (for online help)
- Microsoft Excel 2000 or later (for PowerExport feature)
Specifications
Imaging Multi-Mode Readers | |
Data Collection and Analysis Software |