missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Gen5 Microplate Reader and Imager Software
For use with Imaging Multi-Mode Readers.
£15220.00
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
16441925
|
Agilent BioTek
GEN5IPRIME |
Gen5 Image Prime |
£15220.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Offers a unique combination of power and ease-of-use that drives productivity and saves time
- Helps transform raw data and images into meaningful results
- Performs high magnification, intracellular image analysis to basic ELISA
Gather and Analyze Data Quickly and Easily
- Quick start with >150 prepared protocols available in Gen5 and online
- Endpoint, kinetic, well area scanning, spectral scanning read modes
- One-button quick export sends data directly to Excel, no need for multiple steps
- Predefined Take3 Micro-Volume Plate protocols for direct quantification of 2μL samples
Explore, Image and Analyze Intra-and Inter-cellular Activity
- Acquire high quality images from 1.25x to 100x with Lionheart FX and up to 60x with Cytation
- Combine image acquisition steps and multi-mode detection steps to rapidly obtain and analyze phenotypic and quantitative data
- Advanced features including image stitching, z-projection and digital phase contrast
- Import image files from other programs for analysis in Gen5
Imaging: Methods and Processes
- Automated cell counting and analysis with image statistics
- Time-lapse movie making with automated image alignment
- Montage capture, image stitching
- Z-stacking and z-projection
- High speed imaging, image binning for fast kinetics
QC and Validation Tools
- Fully validated at Agilent BioTek
- Gen5 Validation Package provides an efficient step-by-step approach to facilitate internal validation
- Compatible with the comprehensive microplate reader IQ/OQ/PQ (Product Qualification Packages) to help speed instrument qualification
- Gen5 IVD and Gen5 IVD Image+ have QC trending capability, security features and validation protocols for convenience and efficiency
Gen5 Image Prime for Cell Imaging
- Offers powerful image acquisition and analysis tool for use with Lionheart FX Automated Cell Imager and Cytation Cell Imaging MultiÂMode Readers
- Cell counting, subpopulation analysis, advanced analysis features
- Multi-mode and imaging steps combined in single protocol allow hit-picking for imaging efficiency, no need to collect unnecessary data
- Image processing and analysis including Z-projection, image stitching, hitpicking and digital phase contrast, plus advanced features including secondary mask, dynamic thresholding and subpopulation analysis
- Built-in movie maker for acquired kinetic imaging sequences
- Fluorescence, brightfield, color brightfield and phase contrast image acquisition
- Import image files from other programs for analysis in Gen5
GxP Compliance with Gen5 Secure
- For regulated laboratory environments, Gen5 Secure offers functionality to help comply with the requirements of 21 CFR Part 11
- Offers imaging control for Lionheart FX Automated Live Cell Imager and Cytation Cell Imaging Multi-Mode Readers
- Includes image processing and analysis including z-projection, image stitching, hitpicking and digital phase contrast
- Advanced features include secondary mask, dynamic thresholding and subpopulation analysis
- Complete reader control and data analysis for all Agilent BioTek microplate readers
- Includes features designed to help laboratories maintain critical GxP requirements including 21 CFR Part 11
- Electronic signature for data verification, approval, authorization and other customizable events
- Data, protocol and system audit trails track and record actions with customizable comments
- Multi-level user permission assignment with highly granular list of protectable functions
- Multiple groups, with multiple users and permissions are definable
- User lockout and reset, password aging are all customizable
- Email notifications for administrator-defined events
- Single sign-on setup available
- Multiple administrators can be assigned
Computer System Requirements
- 64-bit version of Windows 7 or Windows 10 Professional
- Intel Core i5 (4th generation) Processor or higher
- Intel 8 USB Chipset or higher
- 8 GB RAM or greater
- 128 GB Hard Drive space or greater
- Monitor resolution of 1600 x 900
- Keyboard and mouse
Software (Lionheart FX requires Gen5 3.0 or higher)
- Microsoft Internet Explorer v 9.0 or later (for online Help)
- Microsoft Excel for Quick Export and PowerExport: Gen5 3.0: Excel 2007 (32-bit edition only) or Excel 2010 - 2016 (32- and 64-bit editions); Gen5 2.0: Excel 2007 - 2016 (32-bit editions only)
Connectivity
- USB 3 port for camera
- USB 2 or 3 port for instrument
Specifications
Imaging Multi-Mode Readers |