Learn More
Invitrogen™ GeneArt™ Genomic Cleavage Detection Kit
The GeneArt Genomic Cleavage Detection Kit provides a simple, reliable, and rapid method for the detection of locus-specific double-strand break formation.
Brand: Invitrogen™ A24372
184.80 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes:
- 1 bottle Cell Lysis Buffer
- 1 tube Proteinase K
- 1 tube PCR Supermix
- 1 tube Water
- 1 tube Detection Enzyme
- 1 tube Detection Reaction Buffer
- 1 tube Control Template & Primers
Description
The GeneArt Genomic Cleavage Detection Kit provides a simple, reliable, and rapid method for the detection of locus-specific double-strand break formation. When using genome editing tools to obtain targeted mutations, it is necessary to determine the efficiency at which these nucleases cleave the target sequence, particularly prior to proceeding to the more laborious and expensive processes of cloning and sequencing.
The GeneArt genomic cleavage detection kit contains all of the reagents required prior to gel analysis. A sample of the edited cell population is used as a direct PCR template for amplification with primers specific to the targeted region. The PCR product is then denatured and re-annealed to produce heteroduplex mismatches where double strand breaks have occurred, resulting in indel introduction. These mismatches are recognized and cleaved by the detection enzyme. This cleavage is both easily detectable and quantifiable using gel analysis.
- Easy—no genomic DNA isolation, direct PCR amplification
- Rapid—4 hour total processing time
- Quantative—gel band density directly correlated to target indel introduction
Recommended Storage
Store all at -5 to -30°C
Specifications
Contains: • 1 bottle Cell Lysis Buffer • 1 tube Proteinase K • 1 tube PCR Supermix • 1 tube Water • 1 tube T7E1 Detection Enzyme • 1 tube T7E1 Detection Reaction Buffer • 1 tube Control Template & Primers Store all components at -5° to -30°C. |
|
Liquid | |
Taq Polymerase | |
CRISPR-Cas9, TAL Effector Nuclease | |
20 reactions | |
20 Reactions |
Primer-probe | |
Kit | |
Fast | |
Genomic Cleavage Detection Kit | |
INDEL | |
20 Reactions |
Safety and Handling
- GeneArt Genomic Cleavage Detection Kit
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Poly(oxy-1,2-ethanediyl) .alpha.-[4-(1,1,3,3-tetramethylbutyl)phenyl]-.omega.-hydroxy, Proteinase
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.