Learn More
Invitrogen™ GeneArt™ Gibson Assembly HiFi Cloning Kit, electrocompetent cells
The GeneArt Gibson Assembly HiFi Cloning Kit enables DNA assembly and cloning via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a one-step, 15–60 minute isothermal reaction.
Brand: Invitrogen™ A46626
133.76 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The GeneArt Gibson Assembly HiFi Cloning Kit enables DNA assembly and cloning via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a one-step, 15–60 minute isothermal reaction. DNA fragments of different lengths are uniformly assembled using complementary overlaps between fragments. The inherent flexibility of this approach is suitable for small and large DNA constructs and includes both single and multiple inserts. The resulting products can be used for a variety of downstream applications including transformation, PCR and rolling circle amplification (RCA). The GeneArt Gibson Assembly HiFi Cloning Kit, electrocompetent cells, is a complete kit that includes master mix, positive control, water, and ElectroMAX DH10B electrocompetent E. coli.
Features of the GeneArt Gibson Assembly HiFi Cloning Kit include:
- Simple—seamlessly assemble and clone up to six DNA fragments in a single reaction
- Efficient—high fidelity provides more correct clones than other methods
- Flexible—design guidelines allow assembly into any vector of your choice
- Convenient—available as master mix kits and cloning kits complete with chemical or electrocompetent cells
- Trusted—over 4000 citations in the scientific literature highlighting great success
Seamless assembly of multiple fragments
GeneArt Gibson Assembly HiFi cloning is a simple, one-step process whereby up to six fragments are combined in a proprietary enzymatic mix in order to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). After a 15–60 minute incubation, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells, yielding clones that are ready for analysis the next day. The required 20- to 40-base pair end homology is designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. This special enzyme mix creates a seamless and covalently bound DNA construct providing high efficiency.
Robust method for maximum efficiency
Due to the covalently bound final product, the GeneArt Gibson Assembly HiFi method allows the utilization of chemically competent cells or electrocompetent cells for the highest transformation efficiency, improving the success of finding the right clone, particularly for more challenging constructs. There is no need for restriction enzymes, ligation, or recombination sites, and the method provides a perfectly seamless construct without unwanted extra bases.
Provides great versatility
The GeneArt Gibson Assembly HiFi method provides versatility, can streamline many techniques through the rapid combination, addition, deletion, or exchange of DNA segments, and eliminates the need to subclone, saving time and effort in the cloning workflow.
Specifications
ElectroMAX DH10B | |
Seamless cloning | |
Positive Control | |
Seamless Cloning & Genetic Assembly | |
GENEART™ | |
10 reactions | |
DNA Assembly |
Electrocompetent E. coli | |
Includes master mix, positive control, water, and ElectroMAX DH10B electrocompetent E. coli. |
|
Kit | |
Up to 6 fragments | |
Cloning Kit | |
Dry Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.