missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GeneGel Kits
Precast polyacrylamide gels and and semi-dry buffer strips specifically designed for use with the GenePhor Electrophoresis System
Brand: Cytiva 17-6000-14
Code : GH
Additional Details : Weight : 0.50000kg
Description
- Feature 24-well polyacrylamide gels of 122 x 110 x 0.5 mm in dimension
- Semi-dry buffer system offers exceptional handling convenience
- Silver staining following separation increases sensitivity and provides a long lasting record of the analysis
GeneGel Excel 12.5/24 Kit
- Ready-to-run gels plus electrode system
- For a broad range of DNA applications: RFLP, SSCP, VNTR and RAPD
- Separation range: 50-500 base pairs with resolution of 6 to 8 bp in the 100–200 bp range
- Includes five gels, 12 buffer strips, and instructions
Specifications
Multiwell | |
24-well | |
5 precast GeneGel Excel 12.5/24 gels, 6 Anode and 6 Cathode Buffer strips, 1 manual, these ready-to-run gels, are 12.5% Acrylamide |
Precast Gel | |
RFLP, SSCP, VNTR, Heteroduplex analysis, ARDRA and RAPD | |
1 Each |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Germ cell mutagenicity Category 1
- Carcinogenicity Category 1