Learn More
Thermo Scientific™ GeneJET RNA Purification Kit
Thermo Scientific GeneJET RNA Purification Kit is a simple and efficient system for purification of total RNA from mammalian cultured cells, tissue, human blood cells, bacteria, and yeast.
Brand: Thermo Scientific™ K0731
168.67 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific GeneJET RNA Purification Kit is a simple and efficient system for purification of total RNA from mammalian cultured cells, tissue, human blood cells, bacteria, and yeast.
The kit utilizes a silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious cesium chloride gradients, alcohol precipitation, or toxic phenol-chloroform extractions.
RNA molecules longer than 200 nucleotides can be isolated with the GeneJET RNA Purification Kit in 15 minutes after the lysis step. The high-quality purified RNA can be used in a wide range of downstream applications, including RT-PCR, RT-qPCR, Northern blotting, and other RNA-based analyses.
Highlights
- Universal—can be used for both cell and tissue samples from a wide range of sources: mammalian blood, mammalian cell cultures, mammalian tissues, insect, yeast, and bacteria
- Efficient—high yields of total RNA
- Fast—15 minutes after lysis step
- Pure—an A260/280 ratio > 1.9
- Convenient—spin columns are capped and assembled with collection tubes
Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
- RT-PCR
- RT-qPCR
- Northern blotting
- Nuclease protection assay
Includes
Proteinase K Lysis Buffer, Wash Buffer 1 (concentrated), Wash Buffer 2 (concentrated), Nuclease-free Water, GeneJET RNA Purification Columns (pre-assembled with Collection Tubes), Collection Tubes (2 mL), Collection Tubes (1.5 mL), Detailed Protocol

Specifications
• 600 μL Proteinase K; store at room temperature until opened, -20°C after opening • 40 mL Lysis Buffer; room temperature • 40 mL Wash Buffer 1 (concentrated); room temperature • 23 mL Wash Buffer 2 (concentrated); room temperature • 30 mL Nuclease-free Water; room temperature • 50 GeneJET RNA Spin Columns with Collection Tubes; For better long-term performance, it is recommended to store the purification columns at 2°C-8°C. • 50 Collection Tubes (2 mL); room temperature • 50 Collection Tubes (1.5 mL); room temperature |
|
15 min. (After sample homogenization) | |
Bacteria, Blood, Cells, Tissue, Yeast, Insects, Enzymatic Reactions | |
Not High-throughput Compatible (Manual) | |
50 Preps | |
Room Temperature | |
Up to 160 μg |
Silica Spin Column | |
50–100 μL | |
Total RNA | |
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling | |
Mini | |
Bacteria: ≤109 cells, Blood: 0.5 mL, Cells: ≤107, Tissue and Insects: 30 mg, Yeast: ≤4 x 108 cells, Enzymatic reactions: ≤100 μL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.