Learn More
BTX™ Genepaddles™ Electroporation Electrodes
Genepaddles™ Electroporation Electrodes are paddle-style reusable electrodes suitable for both in vitro and in vivo gene delivery.
Brand: BTX™ 450122
Description
- Compatible with BTX Gemini X2 and ECM-2001 electroporation systems
- Two models to suit the size of the target area
- Gold plated, noninvasive
- Thickness: 1 mm
- Easily cleaned
- Life span is approximately 1500 pulses
- 450122 and 450123 require 45-0203 holder and 45-0216 cables (not included)
Specifications
In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery | |
Gold-plated | |
Genepaddles | |
Rectangular, 3.5 mm | |
3 mm | |
10 μsec. to 99 msec. in PBS | |
0.03 to 0.39 in. | |
1 mm Gap | |
0.19 in. | |
Non-sterile | |
Digital User | |
Holder (45-0203), Cables (45-0216) |
ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 | |
BTX Electroporation | |
200 V | |
0.11 in. | |
200+ pulses when properly maintained | |
1 Ea. | |
1 to 10 mm | |
Gold Plated, Non-invasive Electrodes | |
5 mm | |
Banana Plug to Micrograbber | |
Square Wave |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Intended for Research use only. Not for use in diagnostic or clinical procedures.