missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Genepaddles™ Electroporation Electrodes

Product Code. 15427290
Click to view available options
Product Type:
Genepaddles
Genepaddles Kit
Length (Metric):
3 mm
5 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15427290

Brand: BTX™ 450169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Genepaddles™ Electroporation Electrodes are paddle-style reusable electrodes suitable for both in vitro and in vivo gene delivery.

  • Compatible with BTX Gemini X2 and ECM-2001 electroporation systems
  • Two models to suit the size of the target area
  • Gold plated, noninvasive
  • Thickness: 1 mm
  • Easily cleaned
  • Life span is approximately 1500 pulses
  • 450122 and 450123 require 45-0203 holder and 45-0216 cables (not included)

Specifications

For Use With (Application) In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery
For Use With (Equipment) ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2
Material Gold-plated
Product Line BTX Electroporators
Product Type Genepaddles Kit
Voltage 200 V
Electrode Shape Rectangular
Includes Micrograbber cable, square post cable, banana plug and banana cable, genetrode holder, genepaddle
Length (English) 0.11 in.
Length (Metric) 3 mm
Life Span ∼200+ set of pulses
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Size (English) Gap 0.03 to 0.39 in.
Size (Metric) Gap 1 to 10 mm
Tip Type Gold Plated, 3 x 5 mm
Width (English) 0.19 in.
Width (Metric) 5 mm
Sterility Non-sterile
Connection Type Banana Plug to Micrograbber
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.