missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ General Assay Microplate Lids
Shop All Corning Products

Click to view available options
Plate Compatibility:
96-well Microplates Only (not 2 mL block)
96-well Plate
96/384-well, Half Area Plate
All Microplates
No. per Case:
100
100/Cs.
50/Cs.
Material:
Acetate
Polystyrene
Unit Size:
Pack of 100
Pack of 50
Description
Specifications
Specifications
| Material | Polystyrene |
| Sterility | Sterile |
| Condensation Rings | Yes |
| No. per Case | 100/Cs. |
| No. per Pack | 1/Pk. |
| Product Type | Low Evaporation Lid |
| Plate Compatibility | 96-well Microplates Only (not 2 mL block) |
| Cut-off Corners | Corner Notch |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction