missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ General Assay Microplate Lids

Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Brand: Corning™ 3931
101.14 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description

Specifications
Low evaporation lid with corner notch and condensation rings | |
Sterile | |
50/Cs. | |
Lid | |
Corner Notch |
Polystyrene | |
Yes | |
25/Pk. | |
96-well Microplates Only (not 2 mL block) |
Product Suggestions
Customers who viewed this item also viewed.
COR001 Lifescience Guid
File size: 7 MB
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ General Assay Microplate Lids > Low Evaporation; 50/Cs
Spot an opportunity for improvement?Share a Content Correction