missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Brand: Thermo Scientific™ 75016109
4488.75 GBP valid until 2024-12-31
Use promo code "21988" to get your promotional price.
This item is not returnable.
View return policy
Description
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
Thermo Scientific TX-750 Blood Tube Package | |
TX-750 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters | |
See individual components of package | |
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | |
Yes |
See individual centrifuge, rotor and accessory data | |
Max in TX-750: 148 x 5/7 mL Blood Tubes, 84 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors) | |
Purchase of Thermo Scientific General Purpose Centrifuge required for operation | |
See Manual |