missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ General-Purpose Narrow-Neck Bottles Product Code.: 11577433
Add to basket

Fisherbrand™ General-Purpose Narrow-Neck Bottles

Product Code. 11577433 Shop All Fisherbrand Products
500 mL
Click to view available options
Capacity (Metric):
30 mL
125 mL
250 mL
500 mL
1000 mL

Product Code. 11577433

Add to basket
Add to basket

Use these general purpose bottles for liquid storage and sampling. Fisherbrand™ General-Purpose Narrow-Neck Bottles offer excellent chemical resistance. Bottles can be recycled.

  • Clear general purpose bottles
  • Polypropylene screw closure

Specifications

Capacity (Metric) 500 mL
Color Translucent
For Use With (Application) For liquid storage and sampling
Includes Polypropylene screw closure
Material HDPE
Closure Included Yes
Mouth Wide
Height (Metric) 93 mm
Product Type Narrow Mouth Bottle
Closure Material Polypropylene
Closure Size 24/R4
Diameter (Metric) 73 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ General-Purpose Narrow-Neck Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.