Learn More
Buerkle™ General Purpose Aseptic Polypropylene Sample Box
Buerkle™ General Purpose Aseptic Polypropylene Sample Box features a lit that can be opened with one hand and a 90° angle hinge.
£0.75 - £2.91
Specifications
Product Type Sample Box
Color Translucent
Description Aseptic Sample box
Closure Color Transparent
Autoclavable Autoclavable
View More Specs
Products 3
Product Code Brand Diameter (Metric) Capacity (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Capacity (Metric) Height (Metric) Price Quantity & Availability  
10513713
View Documents
Buerkle™
6206-0060
31 mm 50 mL 78 mm
£0.75
Each
In Stock
to see stock.
Add to basket
10055271
View Documents
Buerkle™
6206-0100
43 mm 90 mL 80 mm
£0.90
Each
In Stock
to see stock.
Add to basket
10376992
View Documents
Buerkle™
6206-0300
75 mm 300 mL 88 mm
£2.91
Each
In Stock
to see stock.
Add to basket
Description
Description
  • PP, transparent
  • With area for labelling and with graduation
  • Liquid/condensate in the lid runs into an absorbing groove integrated in the lid, nothing is lost, nothing can drip
  • The box is completely tight, has no crevices, is therefore easy to fill and can be emptied without residues
  • Produced under aseptic conditions
  • Autoclavable and suitable for deep freezing
Specifications
Specifications
Sample Box
Aseptic Sample box
Autoclavable
Suitable for Use with Foodstuffs
1
Polypropylene
Translucent
Transparent
Polypropylene
Sealing cover
Yes
Sealing Cover with Absorbing Groove
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Buerkle™ General Purpose Aseptic Polypropylene Sample Box