Learn More
3M™ General Purpose Vinyl Tape 764
This vinyl tape is a cost-effective choice for a variety of hazard, safety marking, and color-coding applications when you’re not dealing with tough conditions or the need for a long-term clean removal. Comes individually wrapped and conveniently packaged.
Brand:  3M™ 7100260286
 View more versions of this product
Product Code. 17771689
  • £102.00 / 12 rolls
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for marking applications in light traffic areas, walls and pipes
  • Use for taping in all non-critical applications
  • Maintains vivid color even when exposed to abrasion and solvents
  • Protects parts, equipment and products from abrasions
  • Bundle and temporarily mark parts, machines, and more
  • Easy-to-apply tape stretches around corners for a reliable, tight fit
  • Fast tack and high holding strength
  • Available in wide variety of colors
  • Can be applied by hand or with floor applicator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
33 m x 50 mm
0.12 mm
White
12 Rolls
Rubber-based Adhesive
Vinyl
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ General Purpose Vinyl Tape 764 > General Purpose Vinyl Tape 764; Adhesive Type: Rubber-based Adhesive; Color: White; Dimensions (L x W): 33 m x 50 mm; Thickness: 0.12 mm; Material: Vinyl