Learn More
3M™ General Purpose Vinyl Tape 764i
Used for all non-critical applications, including colour coding, bundling and safety marking. Our non-corrosive, flexible vinyl tape is ideal for protecting parts, equipment and products from abrasions.
Brand:  3M™ 7000048917
 View more versions of this product
Product Code. 17217780
  • £201.00 / 24 rolls
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Use for taping in all non-critical applications
  • Protects parts, equipment and products from abrasions
  • Bundle items, bundle and temporarily mark parts, machines, plus much more
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas. Lots of different colours available such as yellow, red, blue, green, black, white, etc
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mm x 33 m
0.13 mm
Black
33 m
50 mm