missing translation for 'onlineSavingsMsg'
Learn More

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 15406 mPa.s (25 °C), Paragon Scientific™

Certified in strict accordance with ASTM D2162. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation to ensures the lowest uncertainty of measurement thus providing standards that are certified with primary level accuracy.

Brand:  Paragon Scientific N5100

Additional Details : Weight : 0.50000kg

Product Code. 15143438

  • £78.95 / 500mL
Estimated Shipment: 08-08-2024
to see stock.

Description

Description

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Density g/mL in accordance with ASTM D1480
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) & dynamic viscosity (mPa·s) given at all temperatures
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.