missing translation for 'onlineSavingsMsg'
Learn More
Learn More
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 1690 mPa.s (25 °C), Paragon Scientific™
Certified in strict accordance with ASTM D2162. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation to ensures the lowest uncertainty of measurement thus providing standards that are certified with primary level accuracy.
Brand: Paragon Scientific N750
This item is not returnable.
View return policy
Description
- Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in strict accordance with ASTM D2162
- Density g/mL in accordance with ASTM D1480
- For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
- Kinematic (cSt) & dynamic viscosity (mPa·s) given at all temperatures
- Standard Pack Size 500mL
- Fully traceable to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nominal values quoted. Certified Values will vary from batch to batch.
Spot an opportunity for improvement?Share a Content Correction