Learn More
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 30991 mPa.s (25 °C), Paragon Scientific™
Certified in strict accordance with ASTM D2162. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation to ensures the lowest uncertainty of measurement thus providing standards that are certified with primary level accuracy.
Brand: Paragon Scientific N10200
Additional Details : Weight : 0.50000kg
Description
- Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in strict accordance with ASTM D2162
- Density g/mL in accordance with ASTM D1480
- For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
- Kinematic (cSt) & dynamic viscosity (mPa·s) given at all temperatures
- Standard Pack Size 500mL
- Fully traceable to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Nominal values quoted. Certified Values will vary from batch to batch.