missing translation for 'onlineSavingsMsg'
Learn More

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 4599 mPa.s (25 °C), Paragon Scientific™

Certified in strict accordance with ASTM D2162. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation to ensures the lowest uncertainty of measurement thus providing standards that are certified with primary level accuracy.

Brand:  Paragon Scientific S2000

Product Code. 16060370

  • £124.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Density g/mL in accordance with ASTM D1480
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) & dynamic viscosity (mPa·s) given at all temperatures
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Specifications

Specifications

500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 4599 mPa.s (25 °C), Paragon Scientific™ > Quantity: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Nominal values quoted. Certified Values will vary from batch to batch.