missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ General Use pH Electrode With BNC Plug

Use this electrode for many common sample types such as aqueous samples and drinking water. Fisherbrand™ General Use pH Electrodes are ideal for general lab use and include a BNC Plug.

Brand:  Fisherbrand™ 5773504-003B

Product Code. 11749798

  • £199.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • pH range: 0 to 13
  • Temperature range: 0 to 80°C
Specifications

Specifications

12 mm
120 mm
Glass
0 to 13
Standards
1 m
General use
0°C to 80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.