Learn More
Thermo Scientific™ GeneRuler High Range DNA Ladder
Perform fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels.
£97.50 - £388.00
Specifications
Concentration | 6X |
---|---|
Gel Type | Agarose |
Product Type | High Range DNA Ladder |
Ready to Load | No |
Size Range | 10171 to 48502 bp |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10374880
|
Thermo Scientific™
SM1351 |
50 μg |
£97.50
50µg |
In Stock
Log in to see stock. |
|||||
11843993
|
Thermo Scientific™
SM1352 |
5 x 50 μg |
£388.00
Pack of 5 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
Description
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5hour, 0.4%agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol in the Resources.)
The ladder is supplied with 6X DNA Loading Dye for sample DNA.
Highlights
- Ideal for both DNA sizing and approximate quantification
- Ideal for fast sizing of high molecular weight DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
6X | |
High Range DNA Ladder | |
10171 to 48502 bp | |
Sustainable packaging |
Agarose | |
No | |
Agarose Gels | |
GeneRuler |
For Research Use Only. Not for use in diagnostic procedures.