missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 500 ROX™ dye Size Standard

Product Code. p-7138585 Shop All Applied Biosystems Products
Click to view available options
Quantity:
800 Reactions
9,600 injections
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15849746

Brand: Applied Biosystems™ 4310361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

16 ROX™ dye-labeled, Single-stranded DNA fragments

The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.

The GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments by electrophoresis. GeneScan™ 500 ROX™ Size Standard is designed for sizing DNA fragments in the 35-500 bp range and provides 16 single-stranded ROX™ dye labeled fragments of 35, 50, 75, 100, 139, 150, 160, 200, 250, 300, 340, 350, 400, 450, 490, and 500 bases. Each of the DNA fragments is labeled with a proprietary fluorophore, which results in a single peak when run under both denaturing and non-denaturing conditions.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Product Line GeneScan, ROX
Content And Storage This kit contains ROX size standard and loading buffer.
Store the kit at 2°C to 8°C. (Do not freeze).
Description GeneScan™ 500 ROX™ dye Size Standard, ROX™ Label. Use with 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
For Use With (Application) Sequencing
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
Label or Dye ROX
Product Type Dye
Quantity 9,600 injections
Run Time Standard

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.