Learn More
Thermo Scientific™ GENESYS Fiber Optic Probes
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Brand:  Thermo Scientific™ 840-305000
 View more versions of this product
Product Code. 15841148
  • £2960.00 / Each
Explore more special offers
Description
Description
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard Fiber Optic Dip Probe with 10 mm Tip
Standard Fiber Optic Dip Probe with 10 mm Tip
GENESYS 150 and GENESYS 180 Spectrophotometers