Learn More
Thermo Scientific™ GENESYS Fiber Optic Probes
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Brand:  Thermo Scientific™ 840-307400
 View more versions of this product
Product Code. 15861148
  • £815.00 / Each
Explore more special offers
Description
Description
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mm Pathlength Tip for Dip Probe for GENESYS 150 and GENESYS 180 Spectrophotometers
Tip for Dip Probe
GENESYS 150 and GENESYS 180 spectrophotometers
10 mm