missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Genipin
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Melting Point | 121.0°C |
Formula Weight | 226.23 |
InChI Key | AZKVWQKMDGGDSV-BCMRRPTOSA-N |
SMILES | COC(=O)C1=CO[C@@H](O)[C@H]2[C@@H]1CC=C2CO |
IUPAC Name | methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate |
Molecular Weight (g/mol) | 226.23 |
Chemical Name or Material | Genipin |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Signal Word
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P405-Store locked up.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'