missing translation for 'onlineSavingsMsg'
Learn More
Genistein, Tocris Bioscience™ Product Code.: 18702704

Genistein, Tocris Bioscience™

Product Code. 18702704
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18702704

Brand: Tocris Bioscience 1110/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EGFR kinase inhibitor. Also estrogen and PPARγ ligand

Genistein is a phytoestrogen with a wide range of biological actions. Genistein inhibits protein tyrosine kinases including epidermal growth factor receptor kinase, binds to PPARγ and estrogen receptors and acts as an agonist at GPER. Genistein inhibits aggregation and fibrilization of Aβ42 and amylin37 (hiAPP). In Huntington's disease fibroblast cells, Genistein reduces intracellular mutant huntingtin (mHTT) aggregates and induces degradation of mHTT; in a cellular model of Huntington's disease, Genistein corrects the mutant phenotype through stimulation of autophagy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00016952
InChI Key TZBJGXHYKVUXJN-UHFFFAOYSA-N
Synonym genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
PubChem CID 5280961
ChEBI CHEBI:28088
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
SMILES OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Quantity 10 mg
Formula Weight 270.24
Percent Purity >98%
Chemical Name or Material Genistein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Genistein, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.