missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ Genova-Series Spectrophotometer Accessory, DNACON Solution

Parts and accessories to expand analysis capabilities or make testing easier.

Brand:  Jenway™ 99959-77

Code : NEW

Additional Details : Weight : 0.00200kg


Product Code. 15464990

  • £254.83 / 20mL
Estimated Shipment: 05-12-2024
to see stock.

Description

Description

  • Accessories
  • For use with Genova-Series Spectrophotometer
Specifications

Specifications

DNACON Solution
Genova-Series Spectrophotometer
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DNACON

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : myristyltrimethylammonium bromide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers