Learn More
Gibco™ Gentamicin (10 mg/mL)
GREENER_CHOICE
Gentamicin (10 mg/mL)
Brand:  Gibco™ 15710080
 View more versions of this product
Product Code. 11510506
  • £2065.00 / Pack of 20
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Gentamicin sulfate is a water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea. Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria. Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used for the prevention of bacterial contamination on cell cultures. The recommended working concentration ranges from 0.5 to 50μg/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Dual-Site cGMP Manufacturing
Gibco™ Gentamicin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Gentamicin product made in our Grand Island facility (15710-072). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485, cGMP, FDA
Gentamicin, 20 x 100mL, 10mg/mL, 15 to 30°C, Storage Temperature, Antibiotic, Gentamicin Agent, Antibiotic Reagent Type, 24 Months Shelf Life, ISO 13485/cGMP/FDA Certified, Ambient, Fewer Resources Used/Less Waste
Liquid
Mammalian cell culture, prevention of cell culture contamination
15 to 30°C
10 mg/mL
Antibiotic
20 x 100mL