missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin (10 mg/mL)

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

£14.86

Specifications

Content And Storage Storage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Concentration 10 mg/mL
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Antibiotic
View More Specs

Products 1
Product Code Brand Concentration Quantity Price Quantity & Availability  
Product Code Brand Concentration Quantity Price Quantity & Availability  
15820243
View Documents Promotion Details
Gibco™
15710064
10 mg/mL 10 mL
£14.86
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gentamicin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition protein synthesis and death in susceptible bacteria.

 

  • Effective against wide variety of gram-positive and gram-negative bacteria
  • Used for prevention of bacterial contamination on cell cultures
  • Recommended working concentration ranges from 0.5 to 50μg/mL

 

Cell Culture, Mammalian Cell Culture

Specifications

Specifications

Storage conditions: 15-30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Mammalian Cell Culture
Antibiotic
Prevention of Cell Culture Contamination
10 mg/mL
Liquid
Sterile-filtered
Room Temperature
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

Product Identifier
  • Gentamicin Reagent Solution

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : gentamicin sulphate

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.