missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentamycin Sulfate (White Powder), Fisher BioReagents
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
Description
Gentamycin Sulfate inhibits protein synthesis, preventing microbial contamination in tissue culture applications. It is also used to confer antibiotic resistance/sensitivity in molecular biology procedures.
Specifications
Specifications
Type | Gentamicin Sulfate |
Physical Form | Solid |
CAS | 1405-41-0 |
pH | 3.5 to 5.5 |
Melting Point | 218°C |
Ignition Residue | < 1.0% |
Molecular Formula | C21H43N5O7·H2SO4 |
Merck Index | 15, 4427 |
Packaging | Amber Glass |
Potency | > 590μg/mg |
Show More |
Safety and Handling
Product Identifier
- Gentamicin sulfate
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'